accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 115962-35-1

IUPAC Name: (3R)-2-[(tert-butoxy)carbonyl]-1,2,3,4-tetrahydroisoquinoline-3-carboxylate
Synonyms: (3R)-2-(tert-butoxycarbonyl)-3,4-dihydro-1H-isoquinoline-3-carboxylate
Molecular Weight (g/mol): 276.31
Molecular Formula: C15H18NO4
InChi Key: HFPVZPNLMJDJFB-GFCCVEGCSA-M
SMILES: CC(C)(C)OC(=O)N1CC2=CC=CC=C2C[C@@H]1C([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results