accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 1159996-20-9

NFFFONHF

CAS RN 1159996-20-9

IUPAC Name: N-(4-fluorophenyl)-1-[4-(trifluoromethyl)phenyl]-1,2,3,4-tetrahydroisoquinoline-2-carboxamide
Synonyms: N-(4-fluorophenyl)-1-[4-(trifluoromethyl)phenyl]-3,4-dihydro-1H-isoquinoline-2-carboxamide
Molecular Weight (g/mol): 414.4
Molecular Formula: C23H18F4N2O
InChi Key: ADCDUDFEGFKKQH-UHFFFAOYNA-N
SMILES: FC1=CC=C(NC(=O)N2CCC3=CC=CC=C3C2C2=CC=C(C=C2)C(F)(F)F)C=C1
Color 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results