accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 116-54-1

IUPAC Name: methyl 2,2-dichloroacetate
Synonyms: methyl dichloroacetate
Molecular Weight (g/mol): 142.96
Molecular Formula: C3H4Cl2O2
InChi Key: HKMLRUAPIDAGIE-UHFFFAOYSA-N
SMILES: COC(=O)C(Cl)Cl
Molecular Weight (g/mol) 
  • (2)
  • (5)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (3)
  • (2)
  • (5)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)