accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 1160102-28-2

IUPAC Name: 6-bromo-7-fluoro-3,4-dihydro-2H-1,4-benzoxazine
Synonyms: 6-bromo-7-fluoro-3,4-dihydro-2H-1,4-benzoxazine
Molecular Weight (g/mol): 232.05
Molecular Formula: C8H7BrFNO
InChi Key: CTMRDIDZVAZIRX-UHFFFAOYSA-N
SMILES: FC1=C(Br)C=C2NCCOC2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results