accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 116255-48-2

ClClBrONNN

CAS RN 116255-48-2

IUPAC Name: 1-{[4-bromo-2-(2,4-dichlorophenyl)oxolan-2-yl]methyl}-1H-1,2,4-triazole
Synonyms: bromuconazole
Molecular Weight (g/mol): 377.06
Molecular Formula: C13H12BrCl2N3O
InChi Key: HJJVPARKXDDIQD-UHFFFAOYNA-N
SMILES: ClC1=CC=C(C(Cl)=C1)C1(CN2C=NC=N2)CC(Br)CO1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)