accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 116355-84-1

IUPAC Name: 2-[2-({19-amino-6-[(3,4-dicarboxybutanoyl)oxy]-16,18-dihydroxy-5,9-dimethylicosan-7-yl}oxy)-2-oxoethyl]butanedioic acid
Synonyms: fumonisin B2
Molecular Weight (g/mol): 705.84
Molecular Formula: C34H59NO14
InChi Key: UXDPXZQHTDAXOZ-UHFFFAOYNA-N
SMILES: CCCCC(C)C(OC(=O)CC(CC(O)=O)C(O)=O)C(CC(C)CCCCCCC(O)CC(O)C(C)N)OC(=O)CC(CC(O)=O)C(O)=O
Molecular Weight (g/mol) 
  • (4)
  • (1)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
Percent Purity 
  • (3)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results