accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

FNOOFOOH

CAS RN 116465-48-6

IUPAC Name: 2,5-difluoro-4-nitrobenzoic acid
Synonyms: 2,5-difluoro-4-nitrobenzoic acid
Molecular Weight (g/mol): 203.1
Molecular Formula: C7H3F2NO4
InChi Key: GPTNSBLYGCZJQV-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC(F)=C(C=C1F)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results