CAS RN 1171331-39-7
IUPAC Name:
2-amino-N-(2,2,2-trifluoroethyl)acetamide hydrochloride
Synonyms:
2-amino-N-(2,2,2-trifluoroethyl)acetamide hydrochloride
Molecular Weight (g/mol):
192.57
Molecular Formula:
C4H8ClF3N2O
InChi Key:
DBNFKWRZLGVLSH-UHFFFAOYSA-N
SMILES:
Cl.NCC(=O)NCC(F)(F)F
1
–
1
of
1
results
1
2-Amino-N-(2,2,2-trifluoroethyl)acetamide hydrochloride, 97%
CAS: 1171331-39-7 Molecular Formula: C4H8ClF3N2O Molecular Weight (g/mol): 192.566 MDL Number: MFCD09944516 InChI Key: DBNFKWRZLGVLSH-UHFFFAOYSA-N Synonym: 2-amino-n-2,2,2-trifluoroethyl acetamide hydrochloride,2-amino-n-2,2,2-trifluoroethyl acetamide;hydrochloride,c4h7f3n2o.hcl,2-amino-n-2,2,2-trifluoroethyl acetamide hcl PubChem CID: 42913698 IUPAC Name: 2-amino-N-(2,2,2-trifluoroethyl)acetamide;hydrochloride SMILES: C(C(=O)NCC(F)(F)F)N.Cl