CAS RN 1173019-61-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
4-(3',6'-Dimethyl-3'-heptyl)phenol Monoethoxylate-13C6, TRC
CAS: 1173019-61-8 Molecular Formula: C1113C6H28O2 Molecular Weight (g/mol): 270.36 Synonym: 2-[4-(1-Ethyl-1,4-dimethylpentyl)phenoxy]ethanol-13C6 SMILES: OCCO[13C]1=[13CH][13CH]=[13C](C(CCC(C)C)(C)CC)[13CH]=[13CH]1