accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 117342-20-8

CH3OONOOBOHOH

CAS RN 117342-20-8

IUPAC Name: [3-(methoxycarbonyl)-5-nitrophenyl]boronic acid
Synonyms: 3-(methoxycarbonyl)-5-nitrophenylboronic acid
Molecular Weight (g/mol): 224.96
Molecular Formula: C8H8BNO6
InChi Key: CDGIRLKQNJXHBJ-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC(=CC(=C1)[N+]([O-])=O)B(O)O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results