accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3NNNCO

CAS RN 1174064-57-3

IUPAC Name: 4-isocyanato-1-methyl-1H-pyrazole
Synonyms: 4-isocyanato-1-methylpyrazole
Molecular Weight (g/mol): 123.12
Molecular Formula: C5H5N3O
InChi Key: TXMLNGHIFGKEFH-UHFFFAOYSA-N
SMILES: CN1C=C(C=N1)N=C=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results