accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 1174064-68-6

IUPAC Name: tert-butyl N-[5-(dimethylamino)-1-oxo-1-(piperazin-1-yl)pentan-2-yl]carbamate
Synonyms: tert-butyl N-[5-(dimethylamino)-1-oxo-1-(piperazin-1-yl)pentan-2-yl]carbamate
Molecular Weight (g/mol): 328.46
Molecular Formula: C16H32N4O3
InChi Key: CDZDEHVWAXDQAQ-UHFFFAOYNA-N
SMILES: CN(C)CCCC(NC(=O)OC(C)(C)C)C(=O)N1CCNCC1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results