accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 117516-88-8

IUPAC Name: ethyl 2-amino-4-(pyridin-4-yl)thiophene-3-carboxylate
Synonyms: ethyl 2-amino-4-(pyridin-4-yl)thiophene-3-carboxylate
Molecular Weight (g/mol): 248.3
Molecular Formula: C12H12N2O2S
InChi Key: LKLAPQXKZAVVPT-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=C(N)SC=C1C1=CC=NC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results