CAS RN 117570-53-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
R&D Systems™ DMXAA
A Tocris product, part of R&D Systems. mSTING agonist; induces antitumor immunological responses
2
Vadimezan, TRC
CAS: 117570-53-3 Molecular Formula: C17H14O4 Molecular Weight (g/mol): 282.29 Synonym: 2-(5,6-Dimethyl-9-oxo-9H-xanthen-4-yl)acetic Acid,5,6-Dimethyl-9-oxo-9H-xanthen-4-ylacetic Acid,5,6-Dimethylxanthenone-4-acetic Acid,AS 1404,ASA 404,DMXAA,NSC 640488,5,6-Dimethyl-9-oxo-9H-xanthene-4-acetic Acid IUPAC Name: 2-(5,6-dimethyl-9-oxoxanthen-4-yl)acetic acid SMILES: Cc1ccc2C(=O)c3cccc(CC(=O)O)c3Oc2c1C