accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 117591-20-5

IUPAC Name: benzyl N-[(1S)-3-methyl-1-{[(2S)-1-oxohexan-2-yl]carbamoyl}butyl]carbamate
Synonyms: benzyl N-[(1S)-3-methyl-1-{[(2S)-1-oxohexan-2-yl]carbamoyl}butyl]carbamate
Molecular Weight (g/mol): 362.47
Molecular Formula: C20H30N2O4
InChi Key: PGGUOGKHUUUWAF-ROUUACIJSA-N
SMILES: CCCC[C@H](NC(=O)[C@H](CC(C)C)NC(=O)OCC1=CC=CC=C1)C=O
Product Type 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results