accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 117946-91-5

H3COHNNH

CAS RN 117946-91-5

IUPAC Name: N-[2-(2-benzyl-1H-indol-3-yl)ethyl]acetamide
Synonyms: luzindole
Molecular Weight (g/mol): 292.38
Molecular Formula: C19H20N2O
InChi Key: WVVXBPKOIZGVNS-UHFFFAOYSA-N
SMILES: CC(=O)NCCC1=C(CC2=CC=CC=C2)NC2=CC=CC=C12
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (1)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results