accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 118-00-3

IUPAC Name: 2-amino-9-[3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-6,9-dihydro-3H-purin-6-one
Synonyms: guanosine
Molecular Weight (g/mol): 283.24
Molecular Formula: C10H13N5O5
InChi Key: NYHBQMYGNKIUIF-UHFFFAOYNA-N
SMILES: NC1=NC(=O)C2=C(N1)N(C=N2)C1OC(CO)C(O)C1O
Quantity 
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
Percent Purity 
  • (4)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (4)
  • (3)
  • (4)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (4)
  • (3)
  • (7)