accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

N(R)(S)(R)NH2CHOHH

CAS RN 118-10-5

IUPAC Name: (S)-[(2R,4S,5R)-5-ethenyl-1-azabicyclo[2.2.2]octan-2-yl](quinolin-4-yl)methanol
Synonyms: (+)-cinconine cinchonine (substance) cinchonan-9-ol, (9S)- d-cinchonine (9S)-cinchonan-9-ol
Molecular Weight (g/mol): 294.4
Molecular Formula: C19H22N2O
InChi Key: KMPWYEUPVWOPIM-QAMTZSDWSA-N
SMILES: O[C@H]([C@H]1C[C@@H]2CCN1C[C@@H]2C=C)C1=C2C=CC=CC2=NC=C1
Molecular Weight (g/mol) 
  • (2)
  • (5)
Quantity 
  • (1)
  • (2)
  • (3)
  • (1)
Percent Purity 
  • (2)
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (5)
  • (1)
  • (2)
  • (3)
  • (1)
  • (2)
  • (3)
  • (3)