accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 118-58-1

IUPAC Name: benzyl 2-hydroxybenzoate
Synonyms: benzyl salicylate
Molecular Weight (g/mol): 228.25
Molecular Formula: C14H12O3
InChi Key: ZCTQGTTXIYCGGC-UHFFFAOYSA-N
SMILES: OC1=CC=CC=C1C(=O)OCC1=CC=CC=C1
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)