accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 118-71-8

IUPAC Name: 3-hydroxy-2-methyl-4H-pyran-4-one
Synonyms: talmon 2-methyl-3-oxy-gamma-pyrone 2-methylpyromeconic acid 2-methyl-3-hydroxypyrone palatone maltol larixinic acid vetol corps praline maltol (natural)
Molecular Weight (g/mol): 126.11
Molecular Formula: C6H6O3
InChi Key: XPCTZQVDEJYUGT-UHFFFAOYSA-N
SMILES: CC1=C(O)C(=O)C=CO1
Percent Purity 
  • (1)
  • (6)
Quantity 
  • (2)
  • (2)
  • (2)
  • (1)
Boiling Point 
  • (2)
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (4)
  • (2)
  • (1)
  • (1)
  • (6)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (4)
  • (4)