accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3HOO

CAS RN 118-90-1

IUPAC Name: 2-methylbenzoic acid
Synonyms: o-toluic acid
Molecular Weight (g/mol): 136.15
Molecular Formula: C8H8O2
InChi Key: ZWLPBLYKEWSWPD-UHFFFAOYSA-N
SMILES: CC1=CC=CC=C1C(O)=O
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
Boiling Point 
  • (6)
Percent Purity 
  • (3)
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (6)
  • (3)
  • (3)
  • (3)