accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NNO

CAS RN 118000-43-4

IUPAC Name: imidazo[1,2-a]pyridine-2-carbaldehyde
Synonyms: imidazo[1,2-a]pyridine-2-carbaldehyde
Molecular Weight (g/mol): 146.15
Molecular Formula: C8H6N2O
InChi Key: QKKKBOZQZHBENG-UHFFFAOYSA-N
SMILES: O=CC1=CN2C=CC=CC2=N1
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)