accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NFFFF

CAS RN 118078-66-3

IUPAC Name: 2-fluoro-4-(trifluoromethyl)pyridine
Synonyms: 2-fluoro-4-(trifluoromethyl)pyridine
Molecular Weight (g/mol): 165.09
Molecular Formula: C6H3F4N
InChi Key: DFNQBXZKPUBEIX-UHFFFAOYSA-N
SMILES: FC1=NC=CC(=C1)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results