accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3NNOBr

CAS RN 118183-92-9

IUPAC Name: 3-(4-bromophenyl)-5-methyl-1,2,4-oxadiazole
Synonyms: 3-(4-bromophenyl)-5-methyl-1,2,4-oxadiazole
Molecular Weight (g/mol): 239.07
Molecular Formula: C9H7BrN2O
InChi Key: YWBIOYGLRGIJRT-UHFFFAOYSA-N
SMILES: CC1=NC(=NO1)C1=CC=C(Br)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results