accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 1184-58-3

IUPAC Name: chlorodimethylalumane
Synonyms: chlorodimethylalumane
Molecular Weight (g/mol): 92.5
Molecular Formula: C2H6AlCl
InChi Key: JGHYBJVUQGTEEB-UHFFFAOYSA-M
SMILES: C[Al](C)Cl
Molecular Weight (g/mol) 
  • (1)
  • (1)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results