accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NH2O

CAS RN 118468-18-1

IUPAC Name: 2-(4-phenoxyphenyl)ethan-1-amine
Synonyms: 2-(4-phenoxyphenyl)ethanamine
Molecular Weight (g/mol): 213.28
Molecular Formula: C14H15NO
InChi Key: JNHPLGDXCJAUBX-UHFFFAOYSA-N
SMILES: NCCC1=CC=C(OC2=CC=CC=C2)C=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results