accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

ClONHOOClNa

CAS RN 1184986-70-6

IUPAC Name: sodium 5,7-dichloro-4-oxo-1,4-dihydroquinoline-2-carboxylate
Synonyms: sodium 5,7-dichloro-4-oxo-1H-quinoline-2-carboxylate
Molecular Weight (g/mol): 280.04
Molecular Formula: C10H4Cl2NNaO3
InChi Key: VPRPMJHKWHCUFW-UHFFFAOYSA-M
SMILES: [Na+].[O-]C(=O)C1=CC(=O)C2=C(Cl)C=C(Cl)C=C2N1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results