CAS RN 118499-70-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Ipidacrine Hydrochloride Hydrate, TRC
CAS: 118499-70-0 Molecular Formula: C12 H16 N2 . Cl H . H2 O Molecular Weight (g/mol): 242.75 Synonym: 1H-Cyclopenta[b]quinolin-9-amine, 2,3,5,6,7,8-hexahydro-, hydrochloride, hydrate (1:1:1),1H-Cyclopenta[b]quinolin-9-amine, 2,3,5,6,7,8-hexahydro-, monohydrochloride, monohydrate (9CI),Ipidacrine hydrochloride hydrate IUPAC Name: 2,3,5,6,7,8-hexahydro-1H-cyclopenta[b]quinolin-9-amine;hydrate;hydrochloride SMILES: O.Cl.Nc1c2CCCc2nc3CCCCc13