accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3NNOOBr

CAS RN 1185310-49-9

IUPAC Name: 6-bromo-4-methyl-3-nitroquinoline
Synonyms: 6-bromo-4-methyl-3-nitroquinoline
Molecular Weight (g/mol): 267.08
Molecular Formula: C10H7BrN2O2
InChi Key: MYJZQOHLSJFXDS-UHFFFAOYSA-N
SMILES: CC1=C(C=NC2=CC=C(Br)C=C12)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results