accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

BFFFFFFFFFFFFFFFFFFFFH3CNHCH3

CAS RN 118612-00-3

IUPAC Name: N,N-dimethylanilinium; tetrakis(2,3,4,5,6-pentafluorophenyl)boranuide
Synonyms: N,N-dimethylanilinium; tetrakis(2,3,4,5,6-pentafluorophenyl)boranuide
Molecular Weight (g/mol): 801.23
Molecular Formula: C32H12BF20N
InChi Key: BRHZQNMGSKUUMN-UHFFFAOYSA-O
SMILES: C[NH+](C)C1=CC=CC=C1.FC1=C(F)C(F)=C(C(F)=C1F)[B-](C1=C(F)C(F)=C(F)C(F)=C1F)(C1=C(F)C(F)=C(F)C(F)=C1F)C1=C(F)C(F)=C(F)C(F)=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results