accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OO(S)NH2(S)

CAS RN 118758-48-8

IUPAC Name: (2S,3S)-3-phenylpyrrolidin-1-ium-2-carboxylate
Synonyms: (2S,3S)-3-phenylpyrrolidin-1-ium-2-carboxylate
Molecular Weight (g/mol): 191.23
Molecular Formula: C11H13NO2
InChi Key: VDEMEKSASUGYHM-UWVGGRQHSA-N
SMILES: [O-]C(=O)[C@H]1[NH2+]CC[C@H]1C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results