CAS RN 1187594-09-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
3
of
3
results
1
Baricitinib, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2
Baricitinib, TRC
CAS: 1187594-09-7 Molecular Formula: C16 H17 N7 O2 S Molecular Weight (g/mol): 371.42 IUPAC Name: 2-[1-ethylsulfonyl-3-[4-(7H-pyrrolo[2,3-d]pyrimidin-4-yl)pyrazol-1-yl]azetidin-3-yl]acetonitrile SMILES: CCS(=O)(=O)N1CC(CC#N)(C1)n2cc(cn2)c3ncnc4[nH]ccc34
3
R&D Systems™ Baricitinib
A Tocris product, part of R&D Systems. Highly potent JAK1 and JAK2 inhibitor; also inhibits JAK3 and Tyk2