accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3COOCH3NCH3H3C

CAS RN 1188-33-6

IUPAC Name: (diethoxymethyl)dimethylamine
Synonyms: (diethoxymethyl)dimethylamine
Molecular Weight (g/mol): 147.22
Molecular Formula: C7H17NO2
InChi Key: BWKAYBPLDRWMCJ-UHFFFAOYSA-N
SMILES: CCOC(OCC)N(C)C
Molecular Weight (g/mol) 
  • (2)
  • (5)
Quantity 
  • (3)
  • (1)
  • (1)
  • (2)
Boiling Point 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results