CAS RN 1188890-36-9
IUPAC Name:
N-[4-(morpholin-4-yl)butyl]-1-benzofuran-2-carboxamide hydrochloride
Synonyms:
N-[4-(morpholin-4-yl)butyl]-1-benzofuran-2-carboxamide hydrochloride
Molecular Weight (g/mol):
338.83
Molecular Formula:
C17H23ClN2O3
InChi Key:
PIOACXKZWXHBRB-UHFFFAOYSA-N
SMILES:
Cl.O=C(NCCCCN1CCOCC1)C1=CC2=CC=CC=C2O1
1
–
1
of
1
results
1
Promotions Available
Tocris Bioscience™ CL 82198 hydrochloride
Tocris Bioscience™ CL 82198 hydrochloride
CAS: 1188890-36-9 Molecular Formula: C17H23ClN2O3