CAS RN 119-26-6
CAS RN 119-26-6
IUPAC Name:
(2,4-dinitrophenyl)hydrazine
Synonyms:
2,4-dinitrophenylhydrazine
Molecular Weight (g/mol):
198.14
Molecular Formula:
C6H6N4O4
InChi Key:
HORQAOAYAYGIBM-UHFFFAOYSA-N
SMILES:
NNC1=CC=C(C=C1[N+]([O-])=O)[N+]([O-])=O
1
–
1
of
1
results
1
2,4-Dinitrophenylhydrazine, moist solid, contains min. 30% water
CAS: 119-26-6 Molecular Formula: C6H6N4O4 MDL Number: MFCD00007578 InChI Key: HORQAOAYAYGIBM-UHFFFAOYSA-N Synonym: 2,4-dinitrophenyl hydrazine,2,4-dnph,hydrazine, 2,4-dinitrophenyl,1-hydrazino-2,4-dinitrobenzene,dnph,brady's reagent,2,4-dinitrofenylhydrazin,unii-1n39kd7qpj,ccris 3140,2,4-dnp hydrazine PubChem CID: 3772977 ChEBI: CHEBI:66932 IUPAC Name: (2,4-dinitrophenyl)hydrazine