accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 119-51-7

H3CNHOO(E)

CAS RN 119-51-7

IUPAC Name: (2E)-2-(N-hydroxyimino)-1-phenylpropan-1-one
Synonyms: (2E)-2-(N-hydroxyimino)-1-phenylpropan-1-one
Molecular Weight (g/mol): 163.18
Molecular Formula: C9H9NO2
InChi Key: YPINLRNGSGGJJT-JXMROGBWSA-N
SMILES: C\C(=N/O)C(=O)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results