accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 119-52-8

IUPAC Name: 2-hydroxy-1,2-bis(4-methoxyphenyl)ethan-1-one
Synonyms: 2-hydroxy-1,2-bis(4-methoxyphenyl)ethanone
Molecular Weight (g/mol): 272.3
Molecular Formula: C16H16O4
InChi Key: LRRQSCPPOIUNGX-UHFFFAOYNA-N
SMILES: COC1=CC=C(C=C1)C(O)C(=O)C1=CC=C(OC)C=C1
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (2)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results