accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 119-53-9

IUPAC Name: 2-hydroxy-1,2-diphenylethan-1-one
Synonyms: (+-)-benzoin fenyl-α-hydroxybenzylketon benzoin - chemical tincture of benzoin anxixang styrax benzoin gum extract benzoin preparation benzoin extract (resinoid) aerozoin bitter almond oil camphor
Molecular Weight (g/mol): 212.25
Molecular Formula: C14H12O2
InChi Key: ISAOCJYIOMOJEB-UHFFFAOYNA-N
SMILES: OC(C(=O)C1=CC=CC=C1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (4)
  • (6)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (1)
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (4)
  • (6)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (3)
  • (3)
  • (3)
  • (4)
  • (1)
  • (1)
  • (1)