accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 119-64-2

IUPAC Name: 1,2,3,4-tetrahydronaphthalene
Synonyms: tetralin benzocyclohexane epa pesticide chemical code 055901 tetranap tetralina caswell no. 842A tetralene tetrahydronaphthalene
Molecular Weight (g/mol): 132.21
Molecular Formula: C10H12
InChi Key: CXWXQJXEFPUFDZ-UHFFFAOYSA-N
SMILES: C1CCC2=CC=CC=C2C1
Molecular Weight (g/mol) 
  • (3)
  • (13)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (3)
  • (6)
  • (3)
Percent Purity 
  • (2)
  • (3)
  • (3)
  • (3)
Grade 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (4)
  • (1)
  • (3)
  • (3)
  • (3)
  • (3)
  • (13)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (6)
  • (3)
  • (2)
  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)