accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 119-68-6

IUPAC Name: 2-(methylamino)benzoic acid
Synonyms: N-methylanthranilic acid kyselina 2-methylaminobenzoova kyselina N-methylanthranilova N-methyl-2-aminobenzoic acid N-methyl-o-aminobenzoic acid
Molecular Weight (g/mol): 151.17
Molecular Formula: C8H9NO2
InChi Key: WVMBPWMAQDVZCM-UHFFFAOYSA-N
SMILES: CNC1=CC=CC=C1C(O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results