accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OHOSSOHO

CAS RN 119-80-2

IUPAC Name: 2-[(2-carboxyphenyl)disulfanyl]benzoic acid
Synonyms: benzoic acid, 2,2'-dithiobis-
Molecular Weight (g/mol): 306.35
Molecular Formula: C14H10O4S2
InChi Key: LBEMXJWGHIEXRA-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=CC=C1SSC1=CC=CC=C1C(O)=O
Molecular Weight (g/mol) 
  • (2)
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results