accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

ONNNH2

CAS RN 1190380-49-4

IUPAC Name: 1-(oxan-4-yl)-1H-pyrazol-4-amine
Synonyms: 1-(oxan-4-yl)pyrazol-4-amine
Molecular Weight (g/mol): 167.21
Molecular Formula: C8H13N3O
InChi Key: GZZNBQLBGSVOOZ-UHFFFAOYSA-N
SMILES: NC1=CN(N=C1)C1CCOCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results