CAS RN 1191-15-7
IUPAC Name:
bis(2-methylpropyl)alumanyl
Synonyms:
diisobutylaluminium hydride
Molecular Weight (g/mol):
141.21
Molecular Formula:
C8H18Al
InChi Key:
SIPUZPBQZHNSDW-UHFFFAOYSA-N
SMILES:
CC(C)C[Al]CC(C)C
1
–
10
of
10
results
1
Diisobutylaluminium hydride, 1.2M (20 wt%) solution in toluene, AcroSeal™
CAS: 1191-15-7 | C8H19Al | 142.22 g/mol
2
Diisobutylaluminium hydride, 1M solution in hexane, AcroSeal™
CAS: 1191-15-7 | C8H19Al | 142.22 g/mol
3
Diisobutylaluminium hydride, 1.0M solution in heptane, AcroSeal™
CAS: 1191-15-7 | C8H19Al
4
Diisobutylaluminium hydride, 1.1M solution in cyclohexane, AcroSeal™
CAS: 1191-15-7 | C8H19Al
5
Diisobutylaluminum hydride, 1.1M in cyclohexane, packaged under Nitrogen in resealable AcroSeal™ bottles, Thermo Scientific Chemicals
CAS: 1191-15-7 Molecular Formula: C8H18Al Molecular Weight (g/mol): 141.21 InChI Key: SIPUZPBQZHNSDW-UHFFFAOYSA-N IUPAC Name: bis(2-methylpropyl)alumanyl SMILES: CC(C)C[Al]CC(C)C
6
Diisobutylaluminum hydride, 25% w/w in hexane, Thermo Scientific™
CAS: 1191-15-7 MDL Number: MFCD00008928 Synonym: DIBAL-H
7
Diisobutylaluminum hydride, 1M solution in hexane, Thermo Scientific™
CAS: 1191-15-7 MDL Number: MFCD00008928
8
Diisobutylaluminum hydride, 25% w/w in hexane, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific™
CAS: 1191-15-7 MDL Number: MFCD00008928
9
Diisobutylaluminum hydride, 1M solution in hexane, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific™
CAS: 1191-15-7 MDL Number: MFCD00008928