accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3NNH2N

CAS RN 1192-21-8

IUPAC Name: 1-methyl-1H-pyrazol-5-amine
Synonyms: 2-methylpyrazol-3-amine
Molecular Weight (g/mol): 97.12
Molecular Formula: C4H7N3
InChi Key: JESRNIJXVIFVOV-UHFFFAOYSA-N
SMILES: CN1N=CC=C1N
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)