accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 1192-28-5

IUPAC Name: N-cyclopentylidenehydroxylamine
Synonyms: cyclopentanone, oxime
Molecular Weight (g/mol): 99.13
Molecular Formula: C5H9NO
InChi Key: YGNXYFLJZILPEK-UHFFFAOYSA-N
SMILES: ON=C1CCCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results