accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NH2S

CAS RN 1193-02-8

IUPAC Name: (4-aminophenyl)sulfanide
Synonyms: (4-aminophenyl)sulfanide
Molecular Weight (g/mol): 124.18
Molecular Formula: C6H6NS
InChi Key: WCDSVWRUXWCYFN-UHFFFAOYSA-M
SMILES: NC1=CC=C([S-])C=C1
Boiling Point 
  • (2)
  • (3)
Percent Purity 
  • (2)
  • (3)
Physical Form 
  • (3)
  • (2)
Quantity 
  • (1)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)