accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 1193-11-9

IUPAC Name: 2,2,4-trimethyl-1,3-dioxolane
Synonyms: 2,2,4-trimethyl-1,3-dioxolane
Molecular Weight (g/mol): 116.16
Molecular Formula: C6H12O2
InChi Key: ALTFLAPROMVXNX-UHFFFAOYNA-N
SMILES: CC1COC(C)(C)O1
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)