accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

BrH3COO(R)(S)(S)(R)(S)(S)OH(S)(S)H3C(S)(S)H3CNONH2C

CAS RN 119302-91-9

IUPAC Name: 1-[(1R,2S,3aS,3bR,5aS,7S,8S,9aS,9bS,11aS)-1-(acetyloxy)-7-hydroxy-9a,11a-dimethyl-8-(morpholin-4-yl)-hexadecahydro-1H-cyclopenta[a]phenanthren-2-yl]-1-(prop-2-en-1-yl)pyrrolidin-1-ium bromide
Synonyms: rocuronium bromide 罗库溴铵
Molecular Weight (g/mol): 609.69
Molecular Formula: C32H53BrN2O4
InChi Key: OYTJKRAYGYRUJK-FMCCZJBLSA-M
SMILES: [Br-].CC(=O)O[C@H]1[C@H](C[C@H]2[C@@H]3CC[C@H]4C[C@H](O)[C@H](C[C@]4(C)[C@H]3CC[C@]12C)N1CCOCC1)[N+]1(CC=C)CCCC1
Molecular Weight (g/mol) 
  • (4)
  • (1)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (4)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)