accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 1194-02-1

IUPAC Name: 4-fluorobenzonitrile
Synonyms: benzonitrile, 4-fluoro-
Molecular Weight (g/mol): 121.11
Molecular Formula: C7H4FN
InChi Key: AEKVBBNGWBBYLL-UHFFFAOYSA-N
SMILES: FC1=CC=C(C=C1)C#N
Molecular Weight (g/mol) 
  • (6)
  • (4)
Quantity 
  • (1)
  • (1)
  • (3)
  • (3)
  • (1)
Boiling Point 
  • (3)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (3)
  • (6)
  • (4)
  • (1)
  • (1)
  • (3)
  • (3)
  • (1)
  • (3)
  • (2)
  • (2)