CAS RN 1195-32-0
IUPAC Name:
1-methyl-4-(prop-1-en-2-yl)benzene
Synonyms:
P,α-dimethylstyrene
Molecular Weight (g/mol):
132.21
Molecular Formula:
C10H12
InChi Key:
MMSLOZQEMPDGPI-UHFFFAOYSA-N
SMILES:
CC(=C)C1=CC=C(C)C=C1